siRNA Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768310.1 | XP_008766532.1 | 15, 16 | 2234 | ||
| GenBank mRNA | BC169007.1 | - | 15, 16 | 1931 | 7387040 | |
| X07286.1 | - | 15, 16 | 1933 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkca| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768310.1 | XP_008766532.1 | 15, 16 | 2234 | ||
| GenBank mRNA | BC169007.1 | - | 15, 16 | 1931 | 7387040 | |
| X07286.1 | - | 15, 16 | 1933 | |||