siRNA Details
| Gene Symbol: Prph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012633.2 | NP_036765.2 | 8 | 1366 | ||
| XM_006257321.2 | XP_006257383.1 | 7 | 2245 | |||
| XM_006257322.2 | XP_006257384.1 | 8 | 2147 | |||
| XM_008765669.1 | XP_008763891.1 | 7 | 2247 | |||
| GenBank mRNA | AF031878.1 | - | 8 | 1359 | ||
| BC100656.1 | - | 8 | 1366 | 7936222 | ||

Prph
Genomic Map