siRNA Details
| Gene Symbol: Ptgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | S67721.1 | - | 5 | 463 | ||
| U03388.1 | - | 5 | 463 | |||
| U18060.1 | - | 4 | 411 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptgs1| Gene Symbol: Ptgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | S67721.1 | - | 5 | 463 | ||
| U03388.1 | - | 5 | 463 | |||
| U18060.1 | - | 4 | 411 | |||