siRNA Details
| Gene Symbol: Ptgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017043.4 | NP_058739.4 | 9 | 1305 | ||
| XM_006234063.2 | XP_006234125.1 | 9 | 1304 | |||
| GenBank mRNA | BC081816.1 | - | 9 | 1305 | 7129759 | |
| S67721.1 | - | 9 | 1298 | |||
| U03388.1 | - | 9 | 1298 | |||
| U18060.1 | - | 8 | 1246 | |||

Ptgs1
Genomic Map