siRNA Details
| Gene Symbol: Ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012637.2 | NP_036769.1 | 1, 2 | 145 | ||
| XM_006235638.2 | XP_006235700.1 | 1, 2 | 214 | |||
| XM_006235639.2 | XP_006235701.1 | 1, 2 | 215 | |||
| GenBank mRNA | BC081829.1 | - | 1, 2 | 145 | 7135855 | |
| M33962.1 | - | 1, 2 | 187 | |||

Ptpn1
Genomic Map