siRNA Details
| Gene Symbol: Rara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031528.2 | NP_113716.2 | 7 | 1314 | ||
| XM_008767945.1 | XP_008766167.1 | 7 | 1114 | |||
| GenBank mRNA | BC099830.1 | - | 7 | 1314 | 7933868 | |
| U15211.1 | - | 7 | 1320 | |||

Rara
Genomic Map