siRNA Details
| Gene Symbol: Rbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191832.1 | NP_001178761.1 | 4 | 3523 | ||
| GenBank mRNA | X56159.1 | - | 2 | 123 | ||
| X69523.1 | - | 661 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbp3| Gene Symbol: Rbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191832.1 | NP_001178761.1 | 4 | 3523 | ||
| GenBank mRNA | X56159.1 | - | 2 | 123 | ||
| X69523.1 | - | 661 | ||||