siRNA Details
Gene Symbol: Rbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191832.1 | NP_001178761.1 | 2, 3 | 3307 | ||
GenBank mRNA | X69523.1 | - | 445 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191832.1 | NP_001178761.1 | 2, 3 | 3307 | ||
GenBank mRNA | X69523.1 | - | 445 |