siRNA Details
| Gene Symbol: Ren | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012642.4 | NP_036774.4 | 2, 3 | 260 | ||
| GenBank mRNA | BC078878.1 | - | 2, 3 | 260 | 7126969 | |
| J02941.1 | - | 2, 3 | 453 | |||
| S60054.1 | - | 2, 3 | 270 | |||
| X07033.1 | - | 2, 3 | 270 | |||

Ren
Genomic Map