siRNA Details
| Gene Symbol: Scn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012647.1 | NP_036779.1 | 11 | 1654 | ||
| GenBank mRNA | M22254.1 | - | 11 | 1654 | ||
| X03639.1 | - | 11 | 1654 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn2a| Gene Symbol: Scn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012647.1 | NP_036779.1 | 11 | 1654 | ||
| GenBank mRNA | M22254.1 | - | 11 | 1654 | ||
| X03639.1 | - | 11 | 1654 | |||