siRNA Details
| Gene Symbol: Scnn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012648.1 | NP_036780.1 | 2 | 142 | ||
| XM_008759682.1 | XP_008757904.1 | 2 | 177 | |||
| XM_008759683.1 | XP_008757905.1 | 3 | 387 | |||
| XM_008759684.1 | XP_008757906.1 | 2 | 219 | |||
| XM_008759685.1 | XP_008757907.1 | 2 | 177 | |||
| XM_008759686.1 | XP_008757908.1 | 2 | 177 | |||
| GenBank mRNA | EU627516.1 | - | 1 | 87 | ||
| U35174.1 | - | 1 | 97 | |||
| U35175.1 | - | 1 | 97 | |||
| X77932.1 | - | 2 | 159 | |||

Scnn1b
Genomic Map