siRNA Details
| Gene Symbol: Scnn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012648.1 | NP_036780.1 | 11, 12 | 1523 | ||
| XM_008759682.1 | XP_008757904.1 | 11, 12 | 1558 | |||
| XM_008759683.1 | XP_008757905.1 | 12, 13 | 1768 | |||
| XM_008759684.1 | XP_008757906.1 | 11, 12 | 1600 | |||
| XM_008759685.1 | XP_008757907.1 | 11, 12 | 1577 | |||
| XM_008759686.1 | XP_008757908.1 | 11, 12 | 1577 | |||
| GenBank mRNA | EU627516.1 | - | 10, 11 | 1468 | ||
| U35174.1 | - | 10, 11 | 1478 | |||
| U35175.1 | - | 10, 11 | 1478 | |||
| X77932.1 | - | 11, 12 | 1540 | |||

Scnn1b
Genomic Map