siRNA Details
| Gene Symbol: Sftpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270645.1 | NP_001257574.1 | 5 | 504 | ||
| NM_001270647.1 | NP_001257576.1 | 5 | 715 | |||
| NM_017329.2 | NP_059025.2 | 5 | 501 | |||
| GenBank mRNA | BC085353.1 | - | 5 | 504 | 7121530 | |
| CK364490.1 | - | 5 | 726 | |||
| M15754.1 | - | 5 | 486 | |||
| M33201.1 | - | 5 | 498 | |||
| X13176.1 | - | 5 | 489 | |||
| X13177.1 | - | 5 | 498 | |||

Sftpa1
Genomic Map