siRNA Details
| Gene Symbol: Slc4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012651.2 | NP_036783.2 | 3 | 284 | ||
| XM_008767946.1 | XP_008766168.1 | 3 | 290 | |||
| GenBank mRNA | L02943.1 | - | 3 | 284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc4a1| Gene Symbol: Slc4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012651.2 | NP_036783.2 | 3 | 284 | ||
| XM_008767946.1 | XP_008766168.1 | 3 | 290 | |||
| GenBank mRNA | L02943.1 | - | 3 | 284 | ||