siRNA Details
| Gene Symbol: Slc4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017049.1 | NP_058745.1 | 4 | 566 | ||
| GenBank mRNA | J05167.1 | - | 4 | 566 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc4a3| Gene Symbol: Slc4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017049.1 | NP_058745.1 | 4 | 566 | ||
| GenBank mRNA | J05167.1 | - | 4 | 566 | ||