siRNA Details
| Gene Symbol: Slc9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012652.1 | NP_036784.1 | 3 | 1730 | ||
| GenBank mRNA | M85299.1 | - | 3 | 1730 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9a1| Gene Symbol: Slc9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012652.1 | NP_036784.1 | 3 | 1730 | ||
| GenBank mRNA | M85299.1 | - | 3 | 1730 | ||