siRNA Details
| Gene Symbol: Sod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017050.1 | NP_058746.1 | 4, 5 | 455 | ||
| GenBank mRNA | BC082800.1 | - | 4, 5 | 435 | 7304983 | |
| FQ209495.1 | - | 4, 5 | 456 | |||
| FQ209941.1 | - | 4, 5 | 451 | |||
| FQ210282.1 | - | 4, 5 | 456 | |||
| FQ220715.1 | - | 4, 5 | 455 | |||
| M21060.1 | - | 4, 5 | 358 | |||
| M25157.1 | - | 4, 5 | 414 | |||
| X05634.1 | - | 4, 5 | 354 | |||
| Y00404.1 | - | 4, 5 | 455 | |||

Sod1
Genomic Map