siRNA Details
| Gene Symbol: Sod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017051.2 | NP_058747.1 | 4, 5 | 609 | ||
| GenBank mRNA | BC070913.1 | - | 4, 5 | 609 | 7104698 | |
| Y00497.1 | - | 4, 5 | 537 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sod2| Gene Symbol: Sod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017051.2 | NP_058747.1 | 4, 5 | 609 | ||
| GenBank mRNA | BC070913.1 | - | 4, 5 | 609 | 7104698 | |
| Y00497.1 | - | 4, 5 | 537 | |||