siRNA Details
| Gene Symbol: Sord | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017052.2 | NP_058748.2 | 5 | 520 | ||
| GenBank mRNA | BC088398.1 | - | 5 | 520 | 7112546 | |
| BC098919.1 | - | 5 | 467 | 7373210 | ||
| BC128707.1 | - | 5 | 496 | 8373758 | ||
| FQ209461.1 | - | 5 | 514 | |||
| FQ210296.1 | - | 5 | 521 | |||
| FQ234001.1 | - | 5 | 409 | |||
| X59037.1 | - | 5 | 460 | |||

Sord
Genomic Map