siRNA Details
| Gene Symbol: Sp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012655.2 | NP_036787.2 | 5 | 1939 | ||
| GenBank mRNA | D12768.1 | - | 5 | 1955 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sp1| Gene Symbol: Sp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012655.2 | NP_036787.2 | 5 | 1939 | ||
| GenBank mRNA | D12768.1 | - | 5 | 1955 | ||