siRNA Details
| Gene Symbol: Agxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270659.1 | NP_001257588.1 | 4 | 345 | ||
| NM_001276706.1 | NP_001263635.1 | 8 | 1006 | |||
| NM_030656.2 | NP_085914.2 | 8 | 1006 | |||
| GenBank mRNA | BC088133.1 | - | 8 | 838 | ||
| FQ228944.1 | - | 4 | 345 | |||
| M35270.1 | - | 8 | 1006 | |||
| X06357.1 | - | 8 | 944 | |||

Agxt
Genomic Map