siRNA Details
| Gene Symbol: Agxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270659.1 | NP_001257588.1 | 5 | 437 | ||
| NM_001276706.1 | NP_001263635.1 | 9 | 1098 | |||
| NM_030656.2 | NP_085914.2 | 9 | 1098 | |||
| GenBank mRNA | BC088133.1 | - | 9 | 930 | ||
| FQ228944.1 | - | 5 | 437 | |||
| M35270.1 | - | 9 | 1098 | |||
| X06357.1 | - | 9 | 1036 | |||

Agxt
Genomic Map