siRNA Details
| Gene Symbol: Tap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032055.4 | NP_114444.2 | 7 | 1422 | ||
| GenBank mRNA | BC101854.1 | - | 7 | 1689 | 7933924 | |
| KC222903.1 | - | 7 | 1363 | |||
| KC222905.1 | - | 7 | 1180 | |||
| KC222906.1 | - | 7 | 1363 | |||
| Y10230.1 | - | 7 | 1363 | |||
| Y10231.1 | - | 7 | 1363 | |||
| Y10232.1 | - | 7 | 1363 | |||
| Y10233.1 | - | 7 | 1363 | |||
| Y10234.1 | - | 7 | 1363 | |||
| Y10235.1 | - | 7 | 1363 | |||

Tap1
Genomic Map