siRNA Details
| Gene Symbol: Hnf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012669.1 | NP_036801.1 | 1 | 364 | ||
| GenBank mRNA | J03170.1 | - | 1 | 364 | ||
| X54423.1 | - | 1 | 214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnf1a| Gene Symbol: Hnf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012669.1 | NP_036801.1 | 1 | 364 | ||
| GenBank mRNA | J03170.1 | - | 1 | 364 | ||
| X54423.1 | - | 1 | 214 | |||