siRNA Details
| Gene Symbol: Hnf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012669.1 | NP_036801.1 | 3 | 703 | ||
| GenBank mRNA | J03170.1 | - | 3 | 703 | ||
| X53297.1 | - | 2 | 61 | |||
| X54423.1 | - | 3 | 553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnf1a| Gene Symbol: Hnf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012669.1 | NP_036801.1 | 3 | 703 | ||
| GenBank mRNA | J03170.1 | - | 3 | 703 | ||
| X53297.1 | - | 2 | 61 | |||
| X54423.1 | - | 3 | 553 | |||