siRNA Details
| Gene Symbol: Tg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030988.2 | NP_112250.2 | 7 | 884 | ||
| GenBank mRNA | AB035201.1 | - | 7 | 884 | ||
| AB679745.1 | - | 7 | 858 | |||
| AB682738.1 | - | 7 | 858 | |||
| AF221622.1 | - | 7 | 858 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tg| Gene Symbol: Tg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030988.2 | NP_112250.2 | 7 | 884 | ||
| GenBank mRNA | AB035201.1 | - | 7 | 884 | ||
| AB679745.1 | - | 7 | 858 | |||
| AB682738.1 | - | 7 | 858 | |||
| AF221622.1 | - | 7 | 858 | |||