siRNA Details
| Gene Symbol: Hist1h2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022643.1 | NP_072169.1 | 1 | 162 | ||
| GenBank mRNA | M18045.1 | - | 1 | 162 | ||
| M18046.1 | - | 1 | 162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h2ba| Gene Symbol: Hist1h2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022643.1 | NP_072169.1 | 1 | 162 | ||
| GenBank mRNA | M18045.1 | - | 1 | 162 | ||
| M18046.1 | - | 1 | 162 | |||