siRNA Details
| Gene Symbol: Thrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270854.1 | NP_001257783.1 | 11 | 1474 | ||
| NM_012672.3 | NP_036804.2 | 12 | 1582 | |||
| XM_006251709.2 | XP_006251771.1 | 7 | 1495 | |||
| XM_008770500.1 | XP_008768722.1 | 9 | 1386 | |||
| GenBank mRNA | DQ191165.1 | - | 9 | 1329 | ||
| HM043807.1 | - | 8 | 1371 | |||
| J03819.1 | - | 11 | 1470 | |||
| J03933.1 | - | 11 | 1471 | |||

Thrb
Genomic Map