siRNA Details
| Gene Symbol: Tnnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012676.1 | NP_036808.1 | 13 | 752 | ||
| GenBank mRNA | M26051.1 | - | 13 | 752 | ||
| M26052.1 | - | 13 | 772 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnnt2| Gene Symbol: Tnnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012676.1 | NP_036808.1 | 13 | 752 | ||
| GenBank mRNA | M26051.1 | - | 13 | 752 | ||
| M26052.1 | - | 13 | 772 | |||