siRNA Details
| Gene Symbol: Tph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100634.2 | NP_001094104.1 | 2, 3 | 121 | ||
| XM_006229219.1 | XP_006229281.1 | 2, 3 | 160 | |||
| GenBank mRNA | CB765437.1 | - | 2, 3 | 75 | ||
| X53501.1 | - | 2, 3 | 121 | |||

Tph1
Genomic Map