siRNA Details
| Gene Symbol: Tph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100634.2 | NP_001094104.1 | 6 | 551 | ||
| XM_006229218.2 | XP_006229280.1 | 5 | 677 | |||
| XM_006229219.1 | XP_006229281.1 | 6 | 590 | |||
| GenBank mRNA | CB796568.1 | - | 1 | 12 | ||
| CB809951.1 | - | 2 | 114 | |||
| X53501.1 | - | 6 | 551 | |||

Tph1
Genomic Map