siRNA Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012678.2 | NP_036810.1 | 5 | 510 | ||
GenBank mRNA | FQ225054.1 | - | 521 | |||
FQ225082.1 | - | 521 | ||||
J02780.1 | - | 5 | 513 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012678.2 | NP_036810.1 | 5 | 510 | ||
GenBank mRNA | FQ225054.1 | - | 521 | |||
FQ225082.1 | - | 521 | ||||
J02780.1 | - | 5 | 513 |