siRNA Details
| Gene Symbol: Clu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053021.2 | NP_444180.2 | 4 | 418 | ||
| XM_006252094.2 | XP_006252156.1 | 3 | 296 | |||
| GenBank mRNA | BC061534.1 | - | 3 | 197 | 6886725 | |
| FQ209467.1 | - | 3 | 194 | |||
| FQ209498.1 | - | 3 | 195 | |||
| FQ209623.1 | - | 3 | 193 | |||
| FQ211071.1 | - | 3 | 193 | |||
| FQ213433.1 | - | 3 | 192 | |||
| FQ214098.1 | - | 3 | 238 | |||
| FQ218237.1 | - | 3 | 193 | |||
| FQ218812.1 | - | 3 | 193 | |||
| FQ219030.1 | - | 3 | 193 | |||
| M16975.1 | - | 4 | 418 | |||
| M64723.1 | - | 3 | 183 | |||
| X13231.1 | - | 3 | 182 | |||

Clu
Genomic Map