siRNA Details
| Gene Symbol: Clu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053021.2 | NP_444180.2 | 6 | 1112 | ||
| XM_006252094.2 | XP_006252156.1 | 5 | 990 | |||
| GenBank mRNA | AF314657.1 | - | 2 | 128 | ||
| BC061534.1 | - | 5 | 891 | 6886725 | ||
| FQ218237.1 | - | 5 | 888 | |||
| M16975.1 | - | 6 | 1112 | |||
| M64723.1 | - | 5 | 877 | |||
| X13231.1 | - | 5 | 876 | |||

Clu
Genomic Map