siRNA Details
| Gene Symbol: Ucp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012682.2 | NP_036814.1 | 3, 4 | 715 | ||
| GenBank mRNA | BC088156.1 | - | 3, 4 | 703 | 7380759 | |
| M11814.1 | - | 3, 4 | 713 | |||
| X03894.1 | - | 3, 4 | 625 | |||
| Gene Symbol: LOC100909612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751860.3 | XP_003751908.2 | 1, 2 | 266 | ||

Ucp1
Genomic Map