siRNA Details
| Gene Symbol: Ucp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012682.2 | NP_036814.1 | 2 | 338 | ||
| GenBank mRNA | BC088156.1 | - | 2 | 326 | 7380759 | |
| M11814.1 | - | 2 | 336 | |||
| X03894.1 | - | 2 | 248 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ucp1| Gene Symbol: Ucp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012682.2 | NP_036814.1 | 2 | 338 | ||
| GenBank mRNA | BC088156.1 | - | 2 | 326 | 7380759 | |
| M11814.1 | - | 2 | 336 | |||
| X03894.1 | - | 2 | 248 | |||