siRNA Details
| Gene Symbol: Vipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012685.2 | NP_036817.1 | 4 | 918 | ||
| GenBank mRNA | BC087136.1 | - | 4 | 918 | 7124910 | |
| M86835.1 | - | 4 | 887 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vipr1| Gene Symbol: Vipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012685.2 | NP_036817.1 | 4 | 918 | ||
| GenBank mRNA | BC087136.1 | - | 4 | 918 | 7124910 | |
| M86835.1 | - | 4 | 887 | |||
Set Membership: |
|