siRNA Details
| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031534.2 | NP_113722.2 | 4, 5 | 750 | ||
| XM_006234621.2 | XP_006234683.1 | 4, 5 | 1221 | |||
| GenBank mRNA | X69716.1 | - | 4, 5 | 750 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wt1| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031534.2 | NP_113722.2 | 4, 5 | 750 | ||
| XM_006234621.2 | XP_006234683.1 | 4, 5 | 1221 | |||
| GenBank mRNA | X69716.1 | - | 4, 5 | 750 | ||