siRNA Details
| Gene Symbol: Cckar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012688.3 | NP_036820.1 | 5 | 984 | ||
| XM_008770188.1 | XP_008768410.1 | 6 | 1168 | |||
| GenBank mRNA | M88096.1 | - | 5 | 988 | ||
More Information
Set Membership: |
|

Cckar
Genomic Map