siRNA Details
| Gene Symbol: Esr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012689.1 | NP_036821.1 | 9 | 1799 | ||
| XM_006227832.2 | XP_006227894.1 | 9 | 2399 | |||
| XM_008758703.1 | XP_008756925.1 | 10 | 2447 | |||
| XM_008758704.1 | XP_008756926.1 | 9 | 2371 | |||
| GenBank mRNA | AB477039.1 | - | 9 | 1805 | ||
| KJ746832.1 | - | 9 | 1644 | |||
| KJ746833.1 | - | 10 | 1686 | |||
| X61098.1 | - | 8 | 1663 | |||
| Y00102.1 | - | 9 | 1799 | |||

Esr1
Genomic Map