siRNA Details
| Gene Symbol: Smcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031536.2 | NP_113724.1 | 2 | 390 | ||
| GenBank mRNA | BC078795.1 | - | 2 | 301 | 7134574 | |
| U48702.1 | - | 2 | 396 | |||
| X87883.1 | - | 2 | 382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smcp| Gene Symbol: Smcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031536.2 | NP_113724.1 | 2 | 390 | ||
| GenBank mRNA | BC078795.1 | - | 2 | 301 | 7134574 | |
| U48702.1 | - | 2 | 396 | |||
| X87883.1 | - | 2 | 382 | |||