siRNA Details
| Gene Symbol: Kng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012696.2 | NP_036828.2 | 428 | |||
| GenBank mRNA | BC087028.1 | - | 428 | 7130388 | ||
| FQ219322.1 | - | 309 | ||||
| M11883.1 | - | 198 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kng1| Gene Symbol: Kng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012696.2 | NP_036828.2 | 428 | |||
| GenBank mRNA | BC087028.1 | - | 428 | 7130388 | ||
| FQ219322.1 | - | 309 | ||||
| M11883.1 | - | 198 | ||||