siRNA Details
| Gene Symbol: Dmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005246.1 | NP_001005246.1 | 2 | 143 | ||
| XM_008773252.1 | XP_008771474.1 | 2 | 210 | |||
| XM_008773253.1 | XP_008771475.1 | 2 | 210 | |||
| GenBank mRNA | S38777.1 | - | 2 | 143 | ||
| X65468.1 | - | 2 | 158 | |||
| X69767.1 | - | 2 | 158 | |||

Dmd
Genomic Map