siRNA Details
| Gene Symbol: Rnf112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138613.1 | NP_619516.1 | 8 | 994 | ||
| XM_006246441.2 | XP_006246503.1 | 10 | 2106 | |||
| XM_006246445.2 | XP_006246507.1 | 8 | 997 | |||
| XM_008767774.1 | XP_008765996.1 | 10 | 2534 | |||
| XM_008767775.1 | XP_008765997.1 | 10 | 2531 | |||
| XM_008767776.1 | XP_008765998.1 | 10 | 2243 | |||
| XM_008767777.1 | XP_008765999.1 | 9 | 1179 | |||
| GenBank mRNA | AF054586.1 | - | 8 | 994 | ||
| FQ212014.1 | - | 7 | 907 | |||

Rnf112
Genomic Map