siRNA Details
| Gene Symbol: Yy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173290.1 | NP_775412.1 | 5 | 1189 | ||
| GenBank mRNA | AB080317.1 | - | 5 | 1189 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009537, AK166542, AK076149, BC055899, M74590, and others.

Yy1
Genomic Map