siRNA Details
| Gene Symbol: Plp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030990.2 | NP_112252.1 | 2 | 323 | ||
| XM_006257293.2 | XP_006257355.1 | 5 | 710 | |||
| XM_008773401.1 | XP_008771623.1 | 5 | 709 | |||
| GenBank mRNA | BC093596.1 | - | 1 | 184 | 7314758 | |
| FQ211530.1 | - | 2 | 338 | |||
| FQ212519.1 | - | 1 | 37 | |||
| FQ212680.1 | - | 1 | 37 | |||
| FQ213540.1 | - | 1 | 37 | |||
| FQ213841.1 | - | 1 | 37 | |||
| FQ213943.1 | - | 1 | 37 | |||
| FQ214012.1 | - | 1 | 37 | |||
| FQ214050.1 | - | 1 | 37 | |||
| FQ214153.1 | - | 1 | 37 | |||
| FQ214556.1 | - | 1 | 37 | |||
| M11185.1 | - | 2 | 298 | |||
| X02809.1 | - | 2 | 227 | |||

Plp1
Genomic Map