siRNA Details
| Gene Symbol: Lamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017068.2 | NP_058764.2 | 5 | 769 | ||
| XM_006257486.2 | XP_006257548.1 | 5 | 800 | |||
| GenBank mRNA | BC061990.1 | - | 5 | 769 | 5599402 | |
| D90211.1 | - | 5 | 753 | |||
| M32016.1 | - | 4 | 607 | |||

Lamp2
Genomic Map