siRNA Details
| Gene Symbol: Gcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172091.2 | NP_742088.1 | 3, 4 | 303 | ||
| NM_172092.2 | NP_742089.1 | 3, 4 | 311 | |||
| XM_006247876.2 | XP_006247938.1 | 3, 4 | 417 | |||
| GenBank mRNA | BC081700.1 | - | 3, 4 | 275 | 7192550 | |
| L04796.1 | - | 3, 4 | 273 | |||
| M96674.1 | - | 3, 4 | 313 | |||
| X68692.1 | - | 3, 4 | 273 | |||
More Information
Set Membership: |
|

Gcgr
Genomic Map