siRNA Details
| Gene Symbol: Hspa1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212546.4 | NP_997711.1 | 2 | 1863 | ||
| GenBank mRNA | BC108294.1 | - | 1 | 1725 | 7111250 | |
| BC108297.1 | - | 2 | 1863 | 7115041 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa1l| Gene Symbol: Hspa1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212546.4 | NP_997711.1 | 2 | 1863 | ||
| GenBank mRNA | BC108294.1 | - | 1 | 1725 | 7111250 | |
| BC108297.1 | - | 2 | 1863 | 7115041 | ||