siRNA Details
| Gene Symbol: Psmb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012708.2 | NP_036840.2 | 5 | 533 | ||
| GenBank mRNA | BC091161.1 | - | 5 | 521 | 7366097 | |
| BP500440.1 | - | 5 | 538 | |||
| D10757.1 | - | 5 | 500 | |||
| FQ228039.1 | - | 5 | 507 | |||
| KC222887.1 | - | 5 | 482 | |||
| KC222888.1 | - | 5 | 482 | |||
| KC222889.1 | - | 5 | 482 | |||
| KC222890.1 | - | 5 | 482 | |||
| KC222891.1 | - | 5 | 482 | |||

Psmb9
Genomic Map